Learn More
R&D Systems™ Mouse PDGFRL (NP_081116) VersaClone cDNA
Description
The Mouse PDGFRL (NP_081116) VersaClone cDNA clone contains the complete ORF for PDGFRL, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The PDGFRL insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Informations sur la commande
Ships at ambient temperature
Spécification
Spécification
| Type de produit | cDNA |
| Expression | mPdgfrl |
| Numéro d’adhésion | NP_081116 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formule | cDNA is provided in 10mM Tris-Cl, pH 8.5 |
| Type d’insert | 1141bp |
| Identification génétique (Entrez) | 5157 (Human), 68797 (Mouse), 290771 (Rat) |
| Contenu et stockage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Espèces | Human |
| Quantité | 10 μg |