Learn More
MP Biomedicals™ Aprotinin Protein
Code produit 11420662 Tous les produits MP Biomedicals Produits
Change view
Click to view available options
Quantité:
25 mg
Conditionnement:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
11420662 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 11420662 Fournisseur MP Biomedicals™ Code fournisseur 0219410125
From bovine lung
Protease Inhibitor. Activity: ≥3 TIU/mg. Unit Definition: One trypsin inhibitor unit (TIU), which is equal to 900 kallikrein inhibitor units (KIU) and 8800 Schwert and Takenake units (STU), is the amount of inhibitor per mg which hydrolyzes one μmol of benzoyl-DL-arginine-p-nitroanilide (BAPNA) per minute. One TIU reduces the activity of two trypsin units by 50%. It is reactive against trypsin, chymotrypsin, plasmatic and glandular kininogenases, plasmin, urokinase, clotting factor XIIa, protein C, proteinases of the complement system, and leukocyte and tissue proteinases.
TRUSTED_SUSTAINABILITY
Spécification
À utiliser avec (application) Molecular Biology
Poids moléculaire 6511.23
Nom Aprotinin from Bovine Lung
Plage de pH 5 to 7
Quantité 25 mg
Source Bovine Lung
Conditions de stockage 4°C
Forme Lyophilized