Learn More
Thermo Scientific™ Sensititre™ Cation Adjusted Mueller-Hinton Broth w/ TES
Code produit 15781368 Tous les produits Thermo Scientific Produits
Modifier l'affichage
Click to view available options
Quantité:
100 x 5 ml
10 x 11 ml
100 x 11 ml
Conditionnement:
10 pièces
100 pièces
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
15781368 10 x 11 ml 10 pièces
15351186 100 x 5 ml 100 pièces
15466584 100 x 11 ml 100 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 15781368 Fournisseur Thermo Scientific™ Code fournisseur YT346210
  • ONLINE EXCLUSIVE PRICE
    58.25€
    55.00€ / 10 pièces
    Économisez 3.25€
    5% de réduction
 Article restreint
En stock
Perform susceptibility testing with a full range of supplies, including this broth, to complement and enhance your Thermo Scientific™ Sensititre™ test program.
Perform high-quality susceptibility testing with a complete range of supplies, including Thermo Scientific™ Sensititre™ Cation Adjusted Mueller-Hinton Broth w/ TES, to complement and enhance your Thermo Scientific™ Sensititre™ test program.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Spécification
Description Sensititre™ Cation Adjusted Mueller-Hinton Broth w/ TES
Type de produit Microbiology Media
Type de test Microbial Susceptibility
Nb par paquet 10
Type d’emballage 11 mL/Bottle
Quantité 10 x 11 ml