Learn More
Melford TASSE
Code produit 30018523
Change view
Click to view available options
Quantité:
100 mg
250 mg
1 g
25 g
Conditionnement:
100mg
1g
250mg
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
30018523 250 mg 250mg
30018525 25 g 25g
30018522 100 mg 100mg
30018524 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30018523 Supplier Melford Supplier No. M659000.25
This item is not returnable. View return policy
Substrat fluorescent pourβ -D-Glucuronidase (GUS) codée par le gène gusA isolé d'E.
Substrat éclairé pourβ -D-Glucuronidase (GUS) codée par le gène gusA isolé de E.
Substrat fluorescent pourβ -D-Glucuronidase (GUS) codée par le gène gusA isolé d'E. coli. Le test de fluorescence permet la quantification de l'activité GUS dans les extraits protéiques à une excitation maximale de 365 nm (UV) et une émission maximale de 455 nm (bleu).
Specifications
Quantité 250 mg
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.