Learn More
Mycophenolic Acid-d3, TRC
Code produit 17998419
Change view
Click to view available options
Quantité:
1 mg
10 mg
Conditionnement:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
17998419 1 mg 1mg
17968249 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17998419 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCM8315021MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1185242-90-3
Formule moléculaire C17 2H3 H17 O6
Poids moléculaire (g/mol) 323.36
Synonyme Mycophenolic Acid-(methoxy-D3)
Nom IUPAC (E)-6-[4-hydroxy-7-methyl-3-oxo-6-(trideuteriomethoxy)-1H-2-benzofuran-5-yl]-4-methylhex-4-enoic acid
SMILES [2H]C([2H])([2H])Oc1c(C)c2COC(=O)c2c(O)c1C\C=C(/C)\CCC(=O)O
Specifications
Nom chimique ou matériau Mycophenolic acid-D3
Notes de qualité de la pureté HPLC
Quantité 1 mg
Formule InChI InChI=1S/C17H20O6/c1-9(5-7-13(18)19)4-6-11-15(20)14-12(8-23-17(14)21)10(2)16(11)22-3/h4,20H,5-8H2,1-3H3,(H,18,19)/b9-4+/i3D3
Poids de la formule 323.1448
Pourcentage de pureté >95%