Learn More
N,N'-Diméthyl-3,3'-dithiodipropionamide-d6, TRC
Code produit 30373431
Change view
Click to view available options
Quantité:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
30373431 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30373431 Supplier Toronto Research Chemicals Supplier No. TRCD472707100MG
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Molécules organiques de haute pureté et étalons analytiques, distribués stratégiquement dans le monde entier pour favoriser l'innovation et la réussite commerciale.
TRUSTED_SUSTAINABILITY
Specifications
Nom chimique ou matériau N,N'-Dimethyl-3,3'-dithiodipropionamide-d6
CAS 2374866-79-0
Formule moléculaire C8H10D6N2O2S2
Quantité 100 mg
Synonyme 3,3'-Dithiobis[N-(methyl-d3)propanamide]
Formule InChI InChI=1S/C8H16N2O2S2/c1-9-7(11)3-5-13-14-6-4-8(12)10-2/h3-6H2,1-2H3,(H,9,11)(H,10,12)/i1D3,2D3
SMILES O=C(CCSSCCC(NC([2H])([2H])[2H])=O)NC([2H])([2H])[2H]
Nom IUPAC 3,3'-disulfanediylbis(N-(méthyl-d3)propanamide)
Poids moléculaire (g/mol) 242.39
Poids de la formule 242.1