Learn More
N6-Methyladenosine, MedChemExpress
Code produit 16448976
Encompass_Preferred
Change view
Click to view available options
Quantité:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16448976 100 mg
16432346 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16448976 Fournisseur Medchem Express Code fournisseur HYN0086
MedChemExpress N6-Methyladenosine is the most prevalent internal (non-cap) modification present in the messenger RNA (mRNA) of all higher eukaryotes. N6-Methyladenosine can modifies viral RNAs and has antiviral activities.
N6-Methyladenosine is the most prevalent internal (non-cap) modification present in the messenger RNA (mRNA) of all higher eukaryotes. N6-Methyladenosine can modifies viral RNAs and has antiviral activities.
Spécification
Nom chimique ou matériau N6-Methyladenosine
Forme physique Solid
CAS 1867-73-8
Couleur Blanc
Quantité 100 mg
Notes de qualité de la pureté Research
Formule moléculaire C11H15N5O4
Synonyme 6-Methyladenosine N-Methyladenosine
Informations sur la solubilité DMSO : ≥ 31 mg/mL (110.21 mM)
SMILES CNC1=C(N=CN2[C@H]3[C@H](O)[C@H](O)[C@@H](CO)O3)C2=NC=N1
Poids moléculaire (g/mol) 281.27
Poids de la formule 281.27
Pourcentage de pureté 97.35%
Qualité Research