Learn More
Thermo Scientific™ Offres groupées NanoDrop Ultra Acclaro Pro
Tous les produits Thermo Scientific ProduitsDescription
Les spectrophotomètres et fluorimètres NanoDrop™ Ultra de Thermo Scientific™ constituent la nouvelle génération de détection microvolumétrique. Cet instrument de pointe offre une précision et une sensibilité exceptionnelles pour l’analyse des acides nucléiques et des protéines avec des volumes d’échantillons aussi faibles que 1,0 µl. NanoDrop Ultra est conçu pour fonctionner de manière transparente dans les flux de travail d’un large éventail de domaines, notamment la génomique, la protéomique, la biologie moléculaire, la biotechnologie, le domaine pharmaceutique et la recherche en sciences de la vie. Il offre une précision, une efficacité et une fiabilité inégalées dans un unique appareil compact.
Ces offres groupées comprennent l’achat de l’un des instruments NanoDrop Ultra avec un étalonnage d’usine supplémentaire débloquant le logiciel Acclaro Pro.
Avoid costly setbacks in your workflow with an understanding of sample quality like never before using the Thermo Scientific™ NanoDrop™ Ultra Microvolume UV-Vis Spectrophotometers Fluorometers. Innovative Thermo Scientific™ Acclaro™ Sample Intelligence technology is built into every instrument for improved measurement accuracy and contaminant identification. Quantify and qualify DNA, RNA, and protein samples in seconds with only 1–2 μL, and obtain full spectral data before you decide to use samples in downstream applications. Cuvette compatible models are available for additional flexibility.
Obtain the most complete information about the concentration and quality of your DNA or RNA samples by making absorbance or fluorescence measurements on the same instrument. The NanoDrop Ultra FL and NanoDrop UltraC FL are capable of both microvolume absorbance and fluorescence measurements, thanks to the inclusion of red and blue, solid state light emitting diodes (LEDs).
- High sensitivity — More sensitive than absorbance — Based quantification with the ability to detect down to 0.05 ng/μL dsDNA or RNA user account access.
- Sample prep calculator — Integrated reagent calculator determines amount of dye and buffer needed.
Streamlined workflow
- Fully integrated qPCR recipe calculator — Use the built–in library of kits, or add your favorite, to let the software run the math for you. The qPCR recipe calculator suggests how to prepare the reaction based on your sample's concentration.
- Auto Measure and Auto Blank functions — Multi-sample processing can be streamlined with instant measurements that occur when the pedestal arm is down. These functions can be turned ON or OFF with a single tap on the screen.
Advanced connectivity for data storage, printing and sharing
- Export to the cloud — Export data directly to popular cloud storage services like Microsoft OneDrive, Google Drive, and Thermo Fisher Connect.
- Direct LIMS integration — Send data directly into your LIMS with the assistance of an integrated RESTful API.
- Easy data–to–printer options — Print your results directly from the NanoDrop Ultra instrument via Wi-Fi, Ethernet or USB printer connection; print exported data from your PC using the Nanodrop Ultra PC Control software.
- Data Your Way — Customize and preview the information included in the data file prior to export to ensure it meets any downstream needs.
Accuracy beyond expectations with the Acclaro Pro Software
This bundle includes new versions of the most popular measurement applications, which are unlocked from the factory providing ± 5% Error throughout the absorbance dynamic range.
- Accurate high concentration measurements — Absorbance measurements within 5% error up to 550 Abs (10 mm equivalent), up to 400 mg/mL IgG or 18,000 ng/μL.
- Quick, dilution free process — Take triplicate measurements on the pedestal in under 2 minutes.
- Enhanced calibration – bundled purchases get an additional calibration from the factory to calibrate the instrument for exceptional accuracy within the Acclaro Pro applications.
For research use only; not for diagnostic purposes.
Spécification
Spécification
| Gamme de produits | NanoDrop |
| Type | ND FL et Acclaro Pro |
| À utiliser avec (équipement) | NanoDrop Ultra Spectrophotometers and Fluorometers |
| Comprend | USB flash drive containing the Acclaro Pro software for use with the NanoDrop Ultra Spectrophotometers and Fluorometers NanoDrop FL |