Learn More
STEMCELL Technologies NeuroCult™ Enzymatic Dissociation Kit for Adult CNS Tissue (Mouse and Rat)
Code produit 17108291
Encompass_Preferred
Change view
Click to view available options
Quantité:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
17108291 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 17108291 Fournisseur STEMCELL Technologies Code fournisseur 05715
Kit for enzymatic dissociation of adult mouse and rat CNS tissue
  • NeuroCult™ Enzymatic Dissociation Kit for Adult CNS Tissue (Mouse and Rat) is recommended for the enzymatic digestion and dissociation of adult mouse and rat central nervous system (CNS) tissue
  • NeuroCult™ Enzymatic Dissociation Kit has been optimized so that the entire procedure is reproducible, fast, and yields high cell numbers and viabilities
  • The resulting single-cell suspension is ready for immediate use in downstream applications
Spécification
Type de cellule Neural Stem and Progenitor Cells
Quantité 1 Kit