Learn More
MedChemExpress Neurotensin
Code produit 16409408
Encompass_Preferred
Change view
Click to view available options
Quantité:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16409408 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16409408 Fournisseur MedChemExpress Code fournisseur HYP0234
MedChemExpress Neurotensin, a gut tridecapeptide, acts as a potent cellular mitogen for various colorectal and pancreatic cancers which possess high-affinity neurotensin receptors (NTR).
Spécification
Pureté 97.4%
CAS 39379-15-2
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
À utiliser avec (application) Cancer-Kinase/protease
Poids moléculaire 1672.92 g/mol
Type de produit Peptides
Quantité 10 mg
Conditions d’expédition Room temperature
Solubilité H2O : 33.33 mg/mL (19.92 mM∣Need ultrasonic)