Learn More
MedChemExpress Neurotensin
Code produit 16411888
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16411888 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16411888 Fournisseur MedChemExpress Code fournisseur HYP0234
MedChemExpress Neurotensin, a gut tridecapeptide, acts as a potent cellular mitogen for various colorectal and pancreatic cancers which possess high-affinity neurotensin receptors (NTR).
Spécification
Pureté 97.4%
CAS 39379-15-2
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
À utiliser avec (application) Cancer-Kinase/protease
Poids moléculaire 1672.92 g/mol
Type de produit Peptides
Quantité 5 mg
Conditions d’expédition Room temperature
Solubilité H2O : 33.33 mg/mL (19.92 mM∣Need ultrasonic)