Learn More
NSC 42834, MedChemExpress
Code produit 16479137
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16479137 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16479137 Fournisseur Medchem Express Code fournisseur HY15480
MedChemExpress NSC 42834 (JAK2 Inhibitor V), a novel specific inhibitor of Jak2, inhibits Jak2-V617F and Jak2-WT autophosphorylation in a dose-dependent manner but was not cytotoxic to cells at concentrations that inhibited kinase activity.
NSC 42834 (JAK2 Inhibitor V), a novel specific inhibitor of Jak2, inhibits Jak2-V617F and Jak2-WT autophosphorylation in a dose-dependent manner but was not cytotoxic to cells at concentrations that inhibited kinase activity.
Spécification
Nom chimique ou matériau NSC 42834
Forme physique Oil
CAS 195371-52-9
Couleur Colorless
Quantité 5 mg
Notes de qualité de la pureté Research
Formule moléculaire C23H24N2O
À utiliser avec (application) Cancer-Kinase/protease
Synonyme JAK2 Inhibitor V Z3
Informations sur la solubilité DMSO : ≥ 100 mg/mL (290.32 mM) ∣Ethanol : 100 mg/mL (290.32 mM; Need ultrasonic)
SMILES O=C(C1=CC=CC=C1)C(CCC2=NC=CC=C2)(C)CCC3=NC=CC=C3
Poids moléculaire (g/mol) 344.45
Poids de la formule 344.45
Pourcentage de pureté 96.79%
Qualité Research