Learn More
NSC 601980, MedChemExpress
Code produit 16498796
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16498796 100 mg
16471128 5 mg
16468718 10 mg
16414627 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Code produit 16498796 Fournisseur Medchem Express Code fournisseur HYB1714A
MedChemExpress NSC601980 shows antitumor activity in the yeast screening experiment, which can inhibit cell proliferation in the COLO 205 and HT29 with Log GI 50 of -6.6 and -6.9 respectively.
NSC601980 shows antitumor activity in the yeast screening experiment, which can inhibit cell proliferation in the COLO 205 and HT29 with Log GI 50 of -6.6 and -6.9 respectively.
Spécification
Nom chimique ou matériau NSC 601980
CAS 2070018-27-6
Couleur Red
Notes de qualité de la pureté Research
Formule moléculaire C15H12N4
Quantité 100 mg
Informations sur la solubilité DMSO : 25 mg/mL (100.69 mM; Need ultrasonic and warming)
SMILES CC1=NC2=CC=CC=C2N=C1/N=N/C3=CC=CC=C3
Poids moléculaire (g/mol) 248.28
Poids de la formule 248.28
Pourcentage de pureté 98.11%
Qualité Research
Forme physique Solid
À utiliser avec (application) Cancer-programmed cell death