Learn More
Kern Sohn OIML Class E2 Weight Set, Plastic Case
Description
- Set of weights
- Polished stainless steel
- Milligram weight sets 1 mg to 500 mg are supplied in a removable plastic case
Spécification
Spécification
| Classification | E2 |
| Matériau | Acier inoxydable |
| Matériau du coffret | Plastique |
| Poids (métrique) | 1 à 100 g |