Learn More
WTW™ X6 MANOMETRE OXITOP VERT ET JAUNE JAUNE
Description
- Includes LED display, 2 operating keys
- For use with application to display the momentary value (M) and the stored daily value (S)
- OxiTop™-6 Manual measuring head
- Accessory for BOD self check measurement OxiTop ™ IS
- 3 x measuring head GN
- 3 x measuring head GB
Spécification
Spécification
| Type | Manual BOD Measuring Head |
| À utiliser avec (application) | To display the momentary value (M) and the stored daily value (S) |
| Comprend | Écran LED, 2 touches de commande |