Learn More
p-Nitrophenyl Beta-D-Cellotetraoside, TRC
Code produit 30480715
Change view
Click to view available options
Quantité:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
30480715 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30480715 Fournisseur Toronto Research Chemicals Code fournisseur TRCN50320010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 129411-62-7
Formule moléculaire C30 H45 N O23
Poids moléculaire (g/mol) 787.67
Nom IUPAC (2S,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6S)-6-[(2R,3S,4R,5R,6S)-6-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@H]4[C@H](O)[C@@H](O)[C@H](Oc5ccc(cc5)[N+](=O)[O-])O[C@@H]4CO)O[C@@H]3CO)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
Spécification
Nom chimique ou matériau p-Nitrophenyl beta-D-Cellotetraoside
Quantité 10 mg
Formule InChI InChI=1S/C30H45NO23/c32-5-11-15(36)16(37)20(41)28(48-11)52-25-13(7-34)50-30(22(43)18(25)39)54-26-14(8-35)51-29(23(44)19(26)40)53-24-12(6-33)49-27(21(42)17(24)38)47-10-3-1-9(2-4-10)31(45)46/h1-4,11-30,32-44H,5-8H2/t11-,12-,13-,14-,15-,16+,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28+,29+,30+/m1/s1
Poids de la formule 787.2382