Learn More
p-t-Butylphenyl Diphenyl Phosphate, TRC
Code produit 30005981
Change view
Click to view available options
Quantité:
1 g
10 g
Conditionnement:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30005981 10 g 10g
30005549 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30005981 Fournisseur Toronto Research Chemicals Code fournisseur TRCB69996010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 981-40-8
Formule moléculaire C22H23O4P
Poids moléculaire (g/mol) 382.39
Synonyme p-tert-Butylphenyl Diphenyl Ester Phosphoric Acid, , p-tert-Butylphenol Diphenyl Phosphate, Diphenyl 4-tert-Butylphenyl Phosphate, Diphenyl p-tert-Butylphenyl Phosphate, GZhFK, NSC 2885, p-t-Butylphenyl Diphenyl Phosphate, p-tert-Butylphenyl Diphenyl Phosphate;
Nom IUPAC (4-tert-butylphenyl) diphenyl phosphate
SMILES CC(C)(C)c1ccc(OP(=O)(Oc2ccccc2)Oc3ccccc3)cc1
Spécification
Nom chimique ou matériau p-t-Butylphenyl Diphenyl Phosphate
Notes de qualité de la pureté HPLC
Quantité 10 g
Formule InChI InChI=1S/C22H23O4P/c1-22(2,3)18-14-16-21(17-15-18)26-27(23,24-19-10-6-4-7-11-19)25-20-12-8-5-9-13-20/h4-17H,1-3H3
Poids de la formule 382.1334
Pourcentage de pureté >95%