Learn More
MedChemExpress para-iodoHoechst 33258
Code produit 16462787
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16462787 50 mg
16456856 100 mg
16446708 10 mg
16419267 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Code produit 16462787 Fournisseur MedChemExpress Code fournisseur HY15632
MedChemExpress para-iodoHoechst 33258 is part of a family of blue fluorescent dyes used to stain DNA.
Spécification
Contenu et stockage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Quantité 50 mg
Conditions d’expédition Room temperature
Solubilité DMSO : 20.5 mg/mL (38.36 mM; Need ultrasonic and warming)
Poids moléculaire 534.39 g/mol
CAS 158013-43-5