Learn More
DiscoverX PathHunter™ U2OS PRLR-JAK2 Functional Assay
Description
The PathHunter™ PRLR-JAK2 Functional assay cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting CTKs. This cell line enables assessment of PRLR activation via detection of JAK2 mediated phosphorylation-induced SH2 domain protein recruitment. This product uses EFC technology. The included cell line overexpresses PK-tagged PRLR cytokine receptor, an untagged CTK JAK2 and EA-tagged SH2 domain protein. Upon activation of the cytokine receptor, CTK-based receptor phosphorylation leads to SH2-EA recruitment and production of EFC signal. This stable modified cell line is provided as two vials of cryopreserved cells.
Spécification
Spécification
| Entrée du dosage | CTK Activation |
| Lignée cellulaire | U2OS |
| À utiliser avec (application) | Cell Lines |
| Agoniste primaire | Recombinant Human Prolactin |
| Quantité | 2 flacons |
| Entrée cible | PRLR-JAK2 |
| Numéro d’adhésion | NM_000949 |