Learn More
Intelsius™ PathoShield™
Code produit 15681632
Encompass_Preferred
Modifier l'affichage
Click to view available options
Dimensions:
4
5
8
Dimensions (LxWxH):
191 x 145 x 22 mm
275 x 179 x 43 mm
291 x 191 x 102 mm
Conditionnement:
25 pièces
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Dimensions Dimensions (LxWxH) unitSize
15681632 5 275 x 179 x 43 mm 25 pièces
17378599 8 191 x 145 x 22 mm -
17368599 4 291 x 191 x 102 mm -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 15681632 Fournisseur Intelsius™ Code fournisseur PA025
Il en reste null
Ajouter au panier
Ajouter au panier
The PathoShield™ is a complete shipping solution designed for the transportation of Category A and Category B biological materials. Each system is pre-printed with the required markings and includes all the components required to transport your sample safely.
  • Complete Category B shipping solution includes flexible 95kPa tested compliant secondary, absorbent, labels, and relevant UN markings
  • Range of sizes suitable for shipping through postal and courier networks
  • Suitable for a wide range of primaries including blood vials, swabs and sample containers
  • Supplied flat packed for efficient storage and shipping prior to use
Spécification
Longueur (impériale) extérieure 10.8 in.
Largeur (impériale) extérieure 7.0 in.
Comprend Pouch
Dimensions 5
Hauteur (impériale) extérieure 1.7 in.
Type Sample Transport Packaging
Dimensions (L x l) Pouch: 150 x 190 mm (5.9 x 7.4 in.)
Dimensions (LxWxH) 275 x 179 x 43 mm