Learn More
Pefloxacin Mesylate Dihydrate, TRC
Code produit 30133185
Change view
Click to view available options
Quantité:
5 g
25 g
50 g
Conditionnement:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30133185 50 g 50g
30132797 25 g 25g
17912525 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 30133185 Fournisseur Toronto Research Chemicals Code fournisseur TRCP21831350G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 149676-40-4
Formule moléculaire C17 H20 F N3 O3 . C H4 O3 S . 2 H2 O
Poids moléculaire (g/mol) 465.49
Synonyme Pefloxacin methanesulfonate dihydrate, Pefloxacin mesilate dihydrate, 1-Ethyl-6-fluoro-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid methanesulfonate dihydrate, 1-Ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-3-quinolinecarboxylic acid methanesulfonate hydrate (1:1:2), 3-Quinolinecarboxylic acid, 1-ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-, methanesulfonate, hydrate (1:1:2) (ACI), 3-Quinolinecarboxylic acid, 1-ethyl-6-fluoro-1,4-dihydro-7-(4-methyl-1-piperazinyl)-4-oxo-, monomethanesulfonate, dihydrate (9CI), Pefloxacine mesylate dihydrate, Pefloxacine monomethanesulfonate dihydrate, Pefloxacinium methanesulfonate dihydrate
Nom IUPAC 1-ethyl-6-fluoro-7-(4-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid;methanesulfonic acid;dihydrate
SMILES O.O.CCN1C=C(C(=O)O)C(=O)c2cc(F)c(cc12)N3CCN(C)CC3.CS(=O)(=O)O
Spécification
Nom chimique ou matériau Pefloxacin methane sulfonate dihydrate
Notes de qualité de la pureté HPLC
Quantité 50 g
Formule InChI InChI=1S/C17H20FN3O3.CH4O3S.2H2O/c1-3-20-10-12(17(23)24)16(22)11-8-13(18)15(9-14(11)20)21-6-4-19(2)5-7-21;1-5(2,3)4;;/h8-10H,3-7H2,1-2H3,(H,23,24);1H3,(H,2,3,4);2*1H2
Poids de la formule 465.1581
Pourcentage de pureté >95%