Learn More
R&D Systems™ Pepsin Solution
Product Code. 18298053 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18298053 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18298053 Supplier R&D Systems™ Supplier No. NB90066724
  • ONLINE EXCLUSIVE PRICE
    560.93€
    468.30€ / 50mL
    Économisez 92.63€
    16% de réduction
 Restricted Item
Expédition estimée: 04-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
A Novus product, part of R&D Systems.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 4°C. Do not freeze.
Product Line Novus
Quantity 50 mL
For Use With (Application) Preparation of Working Solutions 1. Solution Pepsin is Ready to Use and Does Not Require Any Preparation. Protocol Recommendations 1. Deparaffinize Tissue Sections and Wash Slides with Wash Buffer. 2. Remove Excess Buffer From Slide Without Letting The Tissue Dry. 3. Depending on The Size of The Tissue Section Add 1-2 Drops of Pepsin Solution. 4. Incubate Slide for 5-10 Minutes (at 37 C. 5. Wash Slide with Wash Buffer and Proceed with Immunostaining.)
Type Pepsin Solution