Learn More
PF-06380101, MedChemExpress
Code produit 16445238
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16445238 10 mg
16437957 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 16445238 Fournisseur Medchem Express Code fournisseur HY12522
MedChemExpress PF-06380101 (Aur0101), an auristatin microtubule inhibitor, is a cytotoxic Dolastatin 10 analogue. PF-06380101 (Aur0101) shows excellent potencies in tumor cell proliferation assays and differential ADME properties when compared to other synthetic auristatin analogues that are used in the preparation of ADCs.
PF-06380101 (Aur0101), an auristatin microtubule inhibitor, is a cytotoxic Dolastatin 10 analogue. PF-06380101 (Aur0101) shows excellent potencies in tumor cell proliferation assays and differential ADME properties when compared to other synthetic auristatin analogues that are used in the preparation of ADCs.
Spécification
Nom chimique ou matériau PF-06380101
Forme physique Solid
CAS 1436391-86-4
Couleur Off-White
Quantité 10 mg
Notes de qualité de la pureté Research
Formule moléculaire C39H62N6O6S
À utiliser avec (application) Cancer-programmed cell death
Synonyme Aur0101 Auristatin-0101
Informations sur la solubilité DMSO : ≥ 65 mg/mL (87.48 mM)
SMILES CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C2=NC=CS2)CC3=CC=CC=C3)=O)OC)=O)OC)N(C([C@H](C(C)C)NC(C(C)(C)N)=O)=O)C
Poids moléculaire (g/mol) 743.01
Poids de la formule 743.01
Pourcentage de pureté 98.33%
Qualité Research