Learn More
Phenoxodiol, MedChemExpress
Code produit 16495818
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité
16495818 10 mg
16478487 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16495818 Supplier Medchem Express Supplier No. HY13721
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
MedChemExpress Phenoxodiol, a synthetic analog of Genestein, activates the mitochondrial caspase system, inhibits XIAP (an apoptosis inhibitor), and sensitizes the cancer cells to Fas-mediated apoptosis. This agent also inhibits DNA topoisomerase II by stabilizing the cleavable complex. Phenoxodiol induces cell cycle arrest in the G1/S phase of the cell cycle and upregulates p21WAF1 via a p53 independent manner.
Phenoxodiol, a synthetic analog of Genestein, activates the mitochondrial caspase system, inhibits XIAP (an apoptosis inhibitor), and sensitizes the cancer cells to Fas-mediated apoptosis. This agent also inhibits DNA topoisomerase II by stabilizing the cleavable complex. Phenoxodiol induces cell cycle arrest in the G1/S phase of the cell cycle and upregulates p21WAF1 via a p53 independent manner.
Specifications
Nom chimique ou matériau Phenoxodiol
CAS 81267-65-4
Couleur Off-White
Notes de qualité de la pureté Research
Formule moléculaire C15H12O3
Quantité 10 mg
Synonyme Idronoxil Dehydroequol Haginin E
Informations sur la solubilité DMSO : ≥ 100 mg/mL (416.23 mM) ∣H2O : 1 mg/mL (4.16 mM; Need ultrasonic)
SMILES OC1=CC=C2C=C(C3=CC=C(O)C=C3)COC2=C1
Poids moléculaire (g/mol) 240.25
Poids de la formule 240.25
Pourcentage de pureté 95.0%
Qualité Research
Forme physique Solid
À utiliser avec (application) Cancer-programmed cell death