Learn More
Pleuromutilin, TRC
Code produit 30135563
Change view
Click to view available options
Quantité:
5 mg
1 g
10 g
Conditionnement:
10g
1g
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30135563 1 g 1g
30006274 5 mg 5mg
30003618 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Code produit 30135563 Fournisseur Toronto Research Chemicals Code fournisseur TRCP6105001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identifiants chimiques
CAS 125-65-5
Formule moléculaire C22 H34 O5
Poids moléculaire (g/mol) 378.5
Synonyme Pleuromutilin, (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5-hydroxy-4,6,9,10-tetramethyl-1-oxodecahydro-3a,9-propano-3aH-cyclopentacycloocten-8-yl Hydroxyacetate, Valnemulin Imp. E (EP), Tiamulin Hydrogen Fumarate Imp. G, Acetic acid, 2-hydroxy-, (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-ethenyldecahydro-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-3a,9-propano-3aH-cyclopentacycloocten-8-yl ester, Acetic acid, hydroxy-, (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-ethenyldecahydro-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-3a,9-propano-3aH-cyclopentacycloocten-8-yl ester (9CI), Acetic acid, hydroxy-, 6-ethenyldecahydro-5-hydroxy-4,6,9,10-tetramethyl-1-oxo-3a,9-propano-3aH-cyclopentacycloocten-8-yl ester, [3aS-(3aα,4β,5α,6α,8β,9α,9aβ,10S*)]-, Glycolic acid, 8-ester with octahydro-5,8-dihydroxy-4,6,9,10-tetramethyl-6-vinyl-3a,9-propano-3aH-cyclopentacycloocten-1(4H)-one (8CI), Pleuromutilin (6CI,7CI), 3a,9-Propano-3aH-cyclopentacyclooctene, acetic acid deriv., (+)-Pleuromutilin, 14-Deoxy-14-[(hydroxyacetyl)oxy]mutilin, Antibiotic A 40104C, BC 757, Drosophilin B, Mutilin 14-glycolate, NSC 121145, Pleuromulin
Nom IUPAC (3aS,4R,5S,6S,8R,9R,9aR,10R)-6-Ethenyl-5-hydroxy-4,6,9,10-tetramethyl-1-oxodecahydro-3a,9-propano-3aH-cyclopentacycloocten-8-yl Hydroxyacetate
SMILES C[C@@H]1CC[C@@]23CCC(=O)[C@H]2[C@]1(C)[C@@H](C[C@@](C)(C=C)[C@@H](O)[C@@H]3C)OC(=O)CO
Spécification
Nom chimique ou matériau Pleuromutilin
Quantité 1 g
Formule InChI InChI=1S/C22H34O5/c1-6-20(4)11-16(27-17(25)12-23)21(5)13(2)7-9-22(14(3)19(20)26)10-8-15(24)18(21)22/h6,13-14,16,18-19,23,26H,1,7-12H2,2-5H3/t13-,14+,16-,18+,19+,20-,21+,22+/m1/s1
Poids de la formule 378.2406