Learn More
Poly(3-hexylthiophene-2,5-diyl), regioregular, low metals, Thermo Scientific™
Marque: Thermo Scientific Alfa Aesar 044745.02
| Quantité | 0.5g |
|---|
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
| 104934-50-1 | |
| 196.352 | |
| DUFPJSOXRHVDOV-UHFFFAOYSA-N | |
| 22950249 | |
| CCCCCCC1=C(SC(=C1)C)C |
| C12H20S | |
| MFCD00217686 | |
| poly 3-hexylthiophene-2,5-diyl, 3-hexyl-2,5-dimethyl-thiophene, poly 3-hexylthiophene-2,5-diyl , regiorandom, poly 3-hexylthiophene-2,5-diyl , regioregular, low metals, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 60000-75000, rr or above, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 65000-75000, rr or above, electronic grade, high purity | |
| 3-hexyl-2,5-dimethylthiophene |
Spécification
| Poly (3-hexylthiophene-2,5-diyl) | |
| 104934-50-1 | |
| MFCD00217686 | |
| Soluble in Chloroform and tetrahydrofuran | |
| DUFPJSOXRHVDOV-UHFFFAOYSA-N | |
| 3-hexyl-2,5-dimethylthiophene | |
| 22950249 |
| Regioregular, low metals | |
| C12H20S | |
| poly 3-hexylthiophene-2,5-diyl, 3-hexyl-2,5-dimethyl-thiophene, poly 3-hexylthiophene-2,5-diyl , regiorandom, poly 3-hexylthiophene-2,5-diyl , regioregular, low metals, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 34000-41000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 50000-70000, rr, electronic grade, high purity, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 60000-75000, rr or above, electronic grade, poly 3-hexylthiophene-2,5-diyl , regioregular, mw 65000-75000, rr or above, electronic grade, high purity | |
| 0.5g | |
| CCCCCCC1=C(SC(=C1)C)C | |
| 196.352 |