Learn More
DiscoverX PrecisION™ hHCN4 Recombinant Stable Cell Line
Description
The PrecisION™ hHCN4 Recombinant Stable Cell Line is a stable, engineered cell line for use in studying HCN4 ion channels, including drug candidates which affect channel function. This cell line enables assessment of ion channel function via detection of electrophysiological signals. The included cell line overexpresses HCN4 ion channels. Upon ligand/agonist induced channel activation, response can be detected via automated or manual electrophysiological techniques. This stable modified cell line is provided as two vials of cryopreserved cells.
Spécification
Spécification
| Entrée du dosage | Ion Channel Activity |
| Lignée cellulaire | CHO-K1 |
| À utiliser avec (application) | Cell Lines |
| Quantité | 2 flacons |
| Entrée cible | HCN4 |
| Numéro d’adhésion | AJ238850 |