Learn More
Unity Lab Services™ Preferred Verification Single-Channel Micropipettor 2 x 5
Product Code. 15346704
Change view
Click to view available options
For Use With (Equipment):
Single-channel Micropipetters
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment)
15346704 Single-channel Micropipetters
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15346704 Supplier Unity Lab Services™ Supplier No. umvp938h
 Restricted Item
Expédition estimée: 26-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
ULS offers a full range of support services and programs for all instruments and supplies, regardless of manufacturer. Maximize your technology investments by anticipating and avoiding problems before they occur.
  • Maintenance is performed on the instrument without prior measurement
  • Disassembly of the lower parts, internal cleaning with disinfectants, lubrication, reassembly, leak test
  • Then a final volumetric check with adjustment if necessary
  • Gravimetric check of the pipettor comprising: volume verification in five weighings, two volumes, without change of cone
  • Production of the verification report with declaration of conformity and labeling
  • Average time: three weeks
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Single-channel Micropipetters
Duration 1 Day
Product Type Preferred Verification
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.