Learn More
Pregnenolone Acetate, TRC
Code produit 30135885
Change view
Click to view available options
Quantité:
2.5 g
25 g
100 g
Conditionnement:
100g
2.50g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
30135885 25 g 25g
30135057 100 g 100g
30008658 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135885 Supplier Toronto Research Chemicals Supplier No. TRCP71221025G
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1778-02-5
Formule moléculaire C23 H34 O3
Poids moléculaire (g/mol) 358.51
Synonyme (3β)-3-(Acetyloxy)-pregn-5-en-20-one, 3β-Acetoxy-5-pregnen-20-one, 3β-Acetoxy-δ5-pregnen-20-one, 3β-Hydroxypregn-5-en-20-one Acetate, 3β-Pregn-5-enol-20-one Acetate, 5-Pregnen-3β-ol-20-one Acetate, 5-Pregnene-3β-acetoxy-20-one, Antofin, Artivis, Enescorb, NSC 64827, Pregenolone Acetate, Pregnenolone Acetate, Pregnenolone-3-acetate, Previsone, Sharmone
Nom IUPAC [(3S,8S,9S,10R,13S,14S,17S)-17-acetyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
SMILES CC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC[C@@H]4C(=O)C)[C@@H]3CC=C2C1
Specifications
Nom chimique ou matériau Pregnenolone Acetate
Quantité 25 g
Formule InChI InChI=1S/C23H34O3/c1-14(24)19-7-8-20-18-6-5-16-13-17(26-15(2)25)9-11-22(16,3)21(18)10-12-23(19,20)4/h5,17-21H,6-13H2,1-4H3/t17-,18-,19+,20-,21-,22-,23+/m0/s1
Poids de la formule 358.2508