Learn More
Unity Lab Services™ Premium Verification Single-Channel Micropipettor 2 x 5
Description
- Verification in the state of the volumes delivered in five weighings, two volumes without change of cone
- Edition of the verification report with declaration of conformity
- Maintenance is carried out on the instrument afterwards
- Disassembly of the lower parts, internal cleaning with disinfectants, lubrication, reassembly, leak test
- A final volumetric control with adjustment if necessary
- Then a verification of the volumes delivered in five weighings, two volumes without change of cone
- Production of the verification report with declaration of conformity and labeling
- Average time: three weeks
Specifications
Specifications
| For Use With (Equipment) | Single-channel Micropipetters |
| Duration | 1 Day |
| Product Type | Premium Verification |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?