Learn More
Lonza™ Milieu de croissance des neurones primaires
Product Code. 11695520
Change view
Click to view available options
Quantité:
1 kit
Unit Size:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité Unit Size
11695520 1 kit 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11695520 Supplier Lonza™ Supplier No. CC4461
  • ONLINE EXCLUSIVE PRICE
    442.00€
    333.06€ / 1 pièces
    Économisez 108.94€
    24% de réduction
 Article restreint
Expédition estimée: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Order Info
Dimensions et type de produit
TRUSTED_SUSTAINABILITY
Specifications
Type de produit Milieu de croissance
Quantité 1 kit
Contenu et stockage Kit contenant un flacon de 200 ml de PNBM et de PNGMTM (quotients simples)