Learn More
MedChemExpress Prolactin Releasing Peptide (12-31), human
Code produit 16492959
Encompass_Preferred
Change view
Click to view available options
Quantité:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16492959 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16492959 Fournisseur MedChemExpress Code fournisseur HYP1530
MedChemExpress Prolactin Releasing Peptide (12-31), human is a fragment of the prolactin releasing peptide (PrRP). Prolactin Releasing Peptide (1-31), human is a high affinity GPR10 ligand that cause the release of the prolactin.
Spécification
CAS 235433-36-0
Contenu et stockage Please store the product under the recommended conditions in the Certificate of Analysis.
À utiliser avec (application) Neuroscience-Neuromodulation
Poids moléculaire 2272.57 g/mol
Type de produit Peptides
Quantité 1 mg
Conditions d’expédition Room temperature