Learn More
MedChemExpress Protease-Activated Receptor-2, amide
Code produit 16451938
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16451938 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16451938 Fournisseur MedChemExpress Code fournisseur HYP0283
MedChemExpress Protease-Activated Receptor-2, amide (SLIGKV-NH2) is a highly potent protease-activated receptor-2 (PAR2) activating peptide.
Spécification
Pureté 98.48%
CAS 190383-13-2
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
À utiliser avec (application) Cancer-programmed cell death
Poids moléculaire 614.78 g/mol
Type de produit Peptides
Quantité 5 mg
Conditions d’expédition Room temperature
Solubilité H2O : 33.33 mg/mL (54.21 mM∣Need ultrasonic)