Learn More
MedChemExpress Protease-Activated Receptor-2, amide
Code produit 16464026
Encompass_Preferred
Change view
Click to view available options
Quantité:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16464026 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16464026 Fournisseur MedChemExpress Code fournisseur HYP0283
MedChemExpress Protease-Activated Receptor-2, amide (SLIGKV-NH2) is a highly potent protease-activated receptor-2 (PAR2) activating peptide.
Spécification
Pureté 98.48%
CAS 190383-13-2
Contenu et stockage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
À utiliser avec (application) Cancer-programmed cell death
Poids moléculaire 614.78 g/mol
Type de produit Peptides
Quantité 25 mg
Conditions d’expédition Room temperature
Solubilité H2O : 33.33 mg/mL (54.21 mM∣Need ultrasonic)