Learn More
PTC299, MedChemExpress
Code produit 16497897
Encompass_Preferred
Change view
Click to view available options
Quantité:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
16497897 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16497897 Supplier Medchem Express Supplier No. HY124593
This item is not returnable. View return policy
MedChemExpress PTC299 is an orally active inhibitor of VEGFA mRNA translation that selectively inhibits VEGF protein synthesis at the post-transcriptional level. PTC299 is also a potent inhibitor of dihydroorotate dehydrogenase (DHODH). PTC299 shows good oral bioavailability and lack of off-target kinase inhibition and myelosuppression. PTC299 can be useful for the research of hematologic malignancies.
PTC299 is an orally active inhibitor of VEGFA mRNA translation that selectively inhibits VEGF protein synthesis at the post-transcriptional level. PTC299 is also a potent inhibitor of dihydroorotate dehydrogenase (DHODH). PTC299 shows good oral bioavailability and lack of off-target kinase inhibition and myelosuppression. PTC299 can be useful for the research of hematologic malignancies.
Specifications
Nom chimique ou matériau PTC299
CAS 1256565-36-2
Couleur Blanc
Notes de qualité de la pureté Research
Formule moléculaire C25H20Cl2N2O3
Quantité 5 mg
Informations sur la solubilité DMSO : 50 mg/mL (106.99 mM; Need ultrasonic)
SMILES O=C(N([C@H]1C2=CC=C(OC)C=C2)CCC3=C1NC4=C3C=C(Cl)C=C4)OC5=CC=C(Cl)C=C5
Poids moléculaire (g/mol) 467.34
Poids de la formule 467.34
Pourcentage de pureté 98.24%
Qualité Research
Forme physique Solid
À utiliser avec (application) Cancer-Kinase/protease