Learn More
Veolia Water Systems™ PURELAB R 15 L/h Purification System
Description
- Used in Flame-AAS, HPLC, Immunocytochemistry, Mammalian and bacterial cell culture, Spectrophotometry, Water analysis
Spécification
Spécification
| Débit | 15 l/h |
| Type | 15 L/hr. Purification System |