Learn More
R&D Systems™ QuantiGlo Immunoassay Control Set 732 for Human IL-2
Code produit 30001954
Modifier l'affichage
Click to view available options
Quantité:
1 de chaque
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
30001954 1 de chaque 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30001954 Fournisseur R&D Systems™ Code fournisseur QC191
  • ONLINE EXCLUSIVE PRICE
    221.93€
    198.45€ / 1 pièces
    Économisez 23.48€
    10% de réduction
 Article restreint
Expédition estimée: 03-12-2026
For use with catalog number Q2000B
QuantiGlo Immunoassay Control Set 732 for Human IL-2 is intended for use as quantitative controls for the determination of cytokine concentrations in biological fluids.
  • Applications: ELISA
  • Analyte: Human IL-2
  • Format: Lyophilized
  • Composition: Recombinant human cytokines in diluted porcine serum with preservatives (<0.1% Sodium Azide)
  • Stability: Unreconstituted controls stable at 2°C to 8°C for at least 6 months; reconstituted stability depends on analyte
  • Research Use: For research use only
TRUSTED_SUSTAINABILITY
Spécification
Contenu et stockage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Substances à analyser détectables IL-2
Type d’échantillon Biological Fluids
Type Immunoassay Control
Identification génétique (Entrez) 3558
Utilisation prévue Control
Gamme de produits R&D Systems
Quantité 1 de chaque
Source Human
Forme Lyophilized