Learn More
R&D Systems™ Quantikine Immunoassay Control Set 822 Human Total PAI-1
Code produit 30001888
Modifier l'affichage
Click to view available options
Quantité:
1 de chaque
Conditionnement:
1 pièce
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
30001888 1 de chaque 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30001888 Fournisseur R&D Systems™ Code fournisseur QC209
  • ONLINE EXCLUSIVE PRICE
    210.00€
    198.45€ / 1 pièces
    Économisez 11.55€
    5% de réduction
 Article restreint
Expédition estimée: 23-09-2026
Ajouter au panier
Ajouter au panier
For use with catalog number DTSE100
For use as quantitative controls for the determination of cytokine concentrations in biological fluids. Concentrations have been assigned using R&D Systems™’ Quantikine™ ELISA kits. Controls are prepared in a buffered protein base with preservatives. They contain recombinant human cytokines at low, medium and high concentrations. Controls are supplied lyophilized.
TRUSTED_SUSTAINABILITY
Spécification
Contenu et stockage Store the unopened product at 2-8°C. Do not use past expiration date.
Identification génétique (Entrez) 5054
Quantité 1 de chaque