Learn More
R&D Systems™ Canine CD45 (XP_005622335) VersaClone cDNA
Description
The Canine CD45 (XP_005622335) VersaClone cDNA clone contains the complete ORF for CD45, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CD45 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Informations sur la commande
Ships at ambient temperature
Spécification
Spécification
| Expression | CD45/PTPRC |
| Numéro d’adhésion | XP_005622335 |
| Identification génétique (Entrez) | 5788 |
| Alias de gène | B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide |
| Contenu et stockage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Espèces | Canine |
| Quantité | 10 μg |
For Reasearch Use Only; Not for Use in Humans