Learn More
R&D Systems™ CellXVivo Human B Cell Expansion Kit
Code produit 30001717
Modifier l'affichage
Click to view available options
Quantité:
1 Kit
Conditionnement:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
30001717 1 Kit 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30001717 Fournisseur R&D Systems™ Code fournisseur CDK005
  • ONLINE EXCLUSIVE PRICE
    531.00€
    501.90€ / 1 kits
    Économisez 29.10€
    5% de réduction
Afficher toutes les promotions
 Article restreint
Expédition estimée: 23-10-2026
Includes
  • The following optimized reagents for the expansion of human B cell populations: B Cell Expander Reagents, Reconstitution Buffers
  • Provides enough reagents for the 3-5 fold expansion of a starting population of 107 human B cells.
For the proliferative expansion of human B cell populations.
TRUSTED_SUSTAINABILITY
Spécification
Contenu et stockage Store the unopened product at 2 - 8° C. Do not use past expiration date.
Quantité 1 Kit