Learn More
R&D Systems™ Methylcellulose Stock Solution
Code produit 18471750
Modifier l'affichage
Click to view available options
Quantité:
100 ml
Conditionnement:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18471750 100 ml 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18471750 Fournisseur R&D Systems™ Code fournisseur HSC001
  • ONLINE EXCLUSIVE PRICE
    234.14€
    208.95€ / 100mL
    Économisez 25.19€
    10% de réduction
 Article restreint
Expédition estimée: 24-11-2026
For the growth and differentiation of human, mouse, and rat hematopoietic stem cells using Iscove's Modified Dulbecco's Media
TRUSTED_SUSTAINABILITY
Spécification
Gamme de produits R&D Systems
Quantité 100 ml
Contenu et stockage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.