Learn More
R&D Systems™ Resazurin
Code produit 18411350
Modifier l'affichage
Click to view available options
Quantité:
100 ml
Conditionnement:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18411350 100 ml 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18411350 Fournisseur R&D Systems™ Code fournisseur AR002
  • ONLINE EXCLUSIVE PRICE
    222.95€
    198.45€ / 100mL
    Économisez 24.50€
    10% de réduction
 Article restreint
En stock
Simple and quantitative method for measuring cell proliferation, viability and cytotoxicity
  • Nontoxic, water soluble, redox-sensitive dye that changes from its blue/non-fluorescent state to pink/highly-fluorescent upon reduction to resorufin by viable cells
  • Fluorescence can be read using 544nm excitation and 590nm emission wavelengths, or absorbance can be read using a spectrophotometer set at 570nm
  • Resazurin may be added at any time during the culture period
  • For measurement of cell proliferation, it is best to add Resazurin during log phase growth
TRUSTED_SUSTAINABILITY
Spécification
Quantité 100 ml
Contenu et stockage Store the unopened product at 2 - 8° C. Protect from light. Do not use past expiration date.
Gamme de produits R&D Systems
Nom du produit
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.