Learn More
R&D Systems™ StemXVivo Xeno-Free Human MSC Expansion Media
Code produit 18482350
Modifier l'affichage
Click to view available options
Quantité:
500 ml
Conditionnement:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18482350 500 ml 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18482350 Fournisseur R&D Systems™ Code fournisseur CCM021
  • ONLINE EXCLUSIVE PRICE
    507.99€
    452.55€ / 500mL
    Économisez 55.44€
    10% de réduction
 Article restreint
Expédition estimée: 24-11-2026
Optimized for the expansion of human mesenchymal stem cells
Mesenchymal stem cells are often expanded in media that contain serum and other components that are of animal origin. The presence of animal-derived factors in media can introduce experimental variability and limit the use of the cultured cells in translational or clinical studies.
  • Maintains MSCs in an undifferentiated state
  • Free of animal components that can introduce variability and pose a safety risk
  • Suitable for translational studies
TRUSTED_SUSTAINABILITY
Spécification
Type de produit Stem Cell Expansion Media
Gamme de produits R&D Systems
Quantité 500 ml
Contenu et stockage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.