Learn More
DiscoverX R-α-Methylhistamine
Code produit 17392489
Encompass
Modifier l'affichage
Click to view available options
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit
17392489 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 17392489 Fournisseur DiscoverX Code fournisseur 921170
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement
R-α-Methylhistamine is an agonist for HRH1-4. Provided as 5mg of dry powder.
(R)(−)-α-Methylhistamine is a potent histamine receptor agonist, particularly H3 (81, 185, and 270 times more potent than histamine for H1, H2, and H3 receptors, respectively). Also binds H4 with Ki=23nM. Inhibits histamine synthesis and release. Provided as 5mg of dry powder.
Spécification
Quantité 5 mg