Learn More
R&D Systems™ Mouse Patched 1/PTCH (NP_032983) VersaClone cDNA
Description
The Mouse Patched 1/PTCH (NP_032983) VersaClone cDNA clone contains the complete ORF for Patched 1/PTCH, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The Patched 1/PTCH insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Informations sur la commande
Ships at ambient temperature
Spécification
Spécification
| Contenu et stockage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantité | 10 μg |
For Reasearch Use Only; Not for Use in Humans