Learn More
rac-Vigabatrin Hydrochloride, TRC
Code produit 30137724
Change view
Click to view available options
Quantité:
5 mg
10 mg
25 mg
Conditionnement:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30137724 25 mg 25mg
18060540 5 mg 5mg
18031250 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30137724 Supplier Toronto Research Chemicals Supplier No. TRCV25301025MG
This item is not returnable. View return policy
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1391054-02-6
Formule moléculaire C6H12ClNO2
Poids moléculaire (g/mol) 165.62
Nom IUPAC 4-aminohex-5-enoic acid;hydrochloride
SMILES Cl.NC(CCC(=O)O)C=C
Specifications
Nom chimique ou matériau rac-Vigabatrin Hydrochloride
Notes de qualité de la pureté HPLC
Quantité 25 mg
Formule InChI InChI=1S/C6H11NO2.ClH/c1-2-5(7)3-4-6(8)9;/h2,5H,1,3-4,7H2,(H,8,9);1H
Poids de la formule 165.0557
Pourcentage de pureté >95%