Learn More
Thermo Scientific™ RadEye PRD4 and PRD-ER4 Personal Radiation Detector
Code produit 17738408 Tous les produits Thermo Scientific Produits
Modifier l'affichage
Click to view available options
Type de détecteur:
1 détecteur de débit de dose faible, 1 détecteur de débit de dose élevé
Description:
Détection personnelle de poche PRD-ER4
Kit pour véhicule PRD-ER4
Conditionnement:
1 pièce
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Type de détecteur Description Conditionnement
17738408 1 détecteur de débit de dose faible, 1 détecteur de débit de dose élevé Kit pour véhicule PRD-ER4 1 pièce
17235506 1 détecteur de débit de dose faible, 1 détecteur de débit de dose élevé Détection personnelle de poche PRD-ER4 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 17738408 Fournisseur Thermo Scientific™ Code fournisseur 425069173
 Article restreint
Cet article nécessite une mise à jour du prix avant d'être commandé.
Demandez un prix pour cet article ici.
missing translation for 'validMainframeMsg'
Thermo Scientific™ RadEye™ PRD4 and PRD-ER4 Personal Radiation Detectors help users detect real radiation threats with high sensitivity while minimizing nuisance alarms that slow screening, response, and decision-making.
Thermo Scientific™ RadEye™ PRD4 and Thermo Scientific™ RadEye™ PRD-ER4 Personal Radiation Detectors help security, emergency response, and radiation safety teams screen people, vehicles, facilities, and materials with high sensitivity, fewer nuisance alarms, and greater confidence in the field. The RadEye PRD4 detector is used for basic screening and interdiction, whereas the RadEye PRD-ER4 detector extends PRD-style screening to higher dose-rate operation capability. Built with natural background rejection (NBR) technology, both the RadEye PRD4 and RadEye PRD-ER4 detectors help distinguish artificial radiation from naturally occurring radioactive material (NORM), thereby reducing time spent adjudicating alarms and improving screening throughput. For missions requiring an added layer of threat awareness, the RadEye PRD4 detector also provides neutron indication capability.
The RadyEye PRD and PRD-ER4 Personal Radiation Detectors enable accurate dose rate measurements across the broader gamma spectrum, providing a greater assurance of personal safety. The neutron indication capability of the RadEye PRD provides an additional layer of detection critical to determining the level of radiation threat.
  • Efficiency in the field
  • Larger display
  • Neutron indication capability
  • 5% improved sensitivity over previous model
  • Linear high dose rate detector
  • Easy information, smarter decisions
  • No retraining or relearning for infrequent users G
  • et results automatically, without the need to press buttons
  • Quickly guides you through next steps after an alarm
  • Simple 4 button design
  • Comprehensive data neatly organized and presented on screen
  • Rugged and practical
  • Can be worn in holster or standard service belt Small and lightweight
  • Long battery life
  • Drop resistant to 1.5m IP65
  • Can be operated in extreme temperatures
TRUSTED_SUSTAINABILITY
Spécification
Description Kit pour véhicule PRD-ER4
Type de produit Personal Radiation Detector
Type de détecteur 1 détecteur de débit de dose faible, 1 détecteur de débit de dose élevé
Comprend RadEye PRD-ER4, Lutetium Test Adapter, Vehicle charging kit, Bluetooth adapter, Software package RadEye.exe, Holster and spare AAA rechargeable batteries