Learn More
R&D Systems™ Rat Cortical Stem Cells (3 x 10e6 cells/vial)
Description
- Have been fully characterized to reduce experimental variation
- Can be expanded as monolayers or neurospheres
- Are free of mycoplasma and microbial contamination
Specifications
Specifications
| Gamme de produits | R&D Systems |
| Quantité | 3 x 10e6 cellules/flacon |