Learn More
R&D Systems™ Rat IL-11 (NP_598203.4) VersaClone cDNA
Spécification
Spécification
| Expression | IL-11 |
| Numéro d’adhésion | NP_598203.4 |
| Identification génétique (Entrez) | 3589 |
| Alias de gène | Adipogenesis inhibitory factor, AGIFoprelvekin, IL-11 Oprelvekin, interleukin 11, interleukin-11 |
| Contenu et stockage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantité | 10 μg |