missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CCL5/RANTES (Mucin Stalk Chimera) Protein
Description
The Recombinant Human CCL5/RANTES (Mucin Stalk Chimera) Protein is derived from NS0. The Recombinant Human CCL5/RANTES (Mucin Stalk Chimera) Protein has been validated for the following applications: Bioactivity.
Tekniske data
Tekniske data
| Formule | Lyophilisé à partir d’une solution filtrée à 0,2 μm dans du PBS avec de la BSA comme protéine porteuse. |
| Identification génétique (Entrez) | 6352 |
| Poids moléculaire | 34 kDa |
| Nom | CCL5/RANTES Mucin Stalk Chimera |
| Quantité | 25 μg |
| Source | Mouse myeloma cell line,NS0-derived human CCL5/RANTES protein, (N-terminus) Human RANTES (Ser24-Ser91) Accession &Num; P13501 GS Human Fractalkine Mucin-like Stalk (Phe103-Gln341) Accession &Num; P78423 KQN 6-His tag (C-terminus) |
| Conditions de stockage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugué | Unconjugated |
| Recombinant | Recombinant |
| Pureté ou qualité | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?